Assay Details
Target Gene Details
Entrez Gene ID: | 55192 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C17 |
Gene Aliases: |
- |
Location: |
Chr.15:40767448-40807478 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC17 | XM_017022401.1 | 12 | 1735 | XP_016877890.1 |
Target Gene Details
Entrez Gene ID: | 2644 |
Gene Name: | GTP cyclohydrolase I feedback regulator |
Gene Aliases: |
GFRP, HsT16933, P35 |
Location: |
Chr.15:40764087-40767713 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCHFR | NM_005258.2 | 3 | 527 | NP_005249.1 |
| AK223430.1 | 3 | 546 | BAD97150.1 | |
| BC112262.1 | 3 | 487 | AAI12263.1 | |
| BC112264.1 | 3 | 487 | AAI12265.1 | |
| BC143904.1 | 3 | 454 |
More Information
Set Membership: |
|

Genomic Map