Assay Details
Target Gene Details
Entrez Gene ID: | 2100 |
Gene Name: | estrogen receptor 2 |
Gene Aliases: |
ER-BETA, ESR-BETA, ESRB, ESTRB, Erb, NR3A2 |
Location: |
Chr.14:64226712-64338631 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESR2 | NM_001040275.1 | 9 | 2198 | NP_001035365.1 |
| NM_001214902.1 | 8 | 2135 | NP_001201831.1 | |
| NM_001291712.1 | 14 | 3458 | NP_001278641.1 | |
| NM_001291723.1 | 9 | 2473 | NP_001278652.1 | |
| NR_073496.1 | 8 | 2334 | ||
| AB006589.1 | 14 | 3005 | BAA31966.1 | |
| AF060555.1 | AAC15234.1 | |||
| BX094834.1 | 1 | 421 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050378 | Chr.14:64219825 - 64236294 on Build GRCh38 | Gain |
|
| nsv564905 | Chr.14:63953695 - 64287643 on Build GRCh38 | Gain |
|
| esv3634746 | Chr.14:64169924 - 64294015 on Build GRCh38 | Loss |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map