Assay Details
Target Gene Details
Entrez Gene ID: | 257106 |
Gene Name: | Rho GTPase activating protein 30 |
Gene Aliases: |
- |
Location: |
Chr.1:161046942-161069971 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP30 | NM_001025598.1 | NP_001020769.1 | ||
| NM_001287600.1 | NP_001274529.1 | |||
| NM_001287602.1 | NP_001274531.1 | |||
| NM_181720.2 | NP_859071.2 | |||
| XM_005245070.2 | XP_005245127.1 | |||
| XM_005245073.3 | XP_005245130.1 | |||
| XM_011509391.2 | XP_011507693.1 | |||
| XM_017000960.1 | XP_016856449.1 | |||
| AK126116.1 | BAC86447.1 | |||
| AK126163.1 | ||||
| AK160366.1 | ||||
| AL832852.1 | ||||
| BC043387.1 | AAH43387.1 | |||
| BC053688.1 | AAH53688.1 | |||
| BX537846.1 | CAD97855.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3221 | Chr.1:161038385 - 161083189 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map