Assay Details
Target Gene Details
Entrez Gene ID: | 926 |
Gene Name: | CD8b molecule |
Gene Aliases: |
CD8B1, LEU2, LY3, LYT3, P37 |
Location: |
Chr.2:86815337-86861924 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD8B | NM_001178100.1 | NP_001171571.1 | ||
| NM_172101.3 | NP_742099.1 | |||
| NM_172102.3 | NP_742100.1 | |||
| NM_172213.3 | NP_757362.1 | |||
| AW296309.1 | ||||
| BQ004743.1 | ||||
| X13445.1 | CAA31796.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10063 | Chr.2:86713865 - 86857237 on Build GRCh38 | Gain |
|
| dgv714e214 | Chr.2:86745573 - 86951144 on Build GRCh38 | Gain |
|
| esv25494 | Chr.2:86745091 - 86946998 on Build GRCh38 | Gain |
|
| nsv458585 | Chr.2:86747195 - 86881088 on Build GRCh38 | Gain |
|
| nsv997982 | Chr.2:86745711 - 86878741 on Build GRCh38 | Gain |
|
| esv3591559 | Chr.2:86764344 - 86832450 on Build GRCh38 | Gain |
|
| dgv3886n100 | Chr.2:86721712 - 86945592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map