Assay Details
Target Gene Details
Entrez Gene ID: | 7809 |
Gene Name: | barttin CLCNK type accessory beta subunit |
Gene Aliases: |
BART, DFNB73 |
Location: |
Chr.1:54998944-55008792 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSND | NM_057176.2 | 3 | 688 | NP_476517.1 |
| AK129999.1 | 3 | 699 | ||
| AY034632.1 | 3 | 688 | AAK57750.1 | |
| BC103898.1 | 3 | 667 | AAI03899.1 | |
| BC103899.1 | 3 | 667 | AAI03900.1 | |
| BC103900.1 | 3 | 667 | AAI03901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv167n100 | Chr.1:54985639 - 55038977 on Build GRCh38 | Gain |
|
| nsv817614 | Chr.1:54580862 - 55735351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map