Assay Details
Target Gene Details
Entrez Gene ID: | 2583 |
Gene Name: | beta-1,4-N-acetyl-galactosaminyltransferase 1 |
Gene Aliases: |
GALGT, GALNACT, GalNAc-T, SPG26 |
Location: |
Chr.12:57623409-57633239 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALNT1 | NM_001276468.1 | 10 | 2078 | NP_001263397.1 |
| NM_001478.4 | 11 | 2570 | NP_001469.1 | |
| XM_005268773.4 | 11 | 2563 | XP_005268830.1 | |
| XM_011538147.2 | 12 | 2581 | XP_011536449.1 | |
| XM_011538148.2 | 12 | 2548 | XP_011536450.1 | |
| XM_017019140.1 | 11 | 2665 | XP_016874629.1 | |
| XM_017019141.1 | 11 | 2377 | XP_016874630.1 | |
| XM_017019142.1 | 11 | 2242 | XP_016874631.1 | |
| AB209460.1 | 10 | 2745 | BAD92697.1 | |
| AK127448.1 | 2 | 2465 | ||
| M83651.1 | 11 | 2161 | AAA35516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map