Assay Details
Target Gene Details
Entrez Gene ID: | 9423 |
Gene Name: | netrin 1 |
Gene Aliases: |
NTN1L |
Location: |
Chr.17:9021542-9244000 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTN1 | NM_004822.2 | 2 | 267 | NP_004813.2 |
| XM_006721595.3 | 2 | 367 | XP_006721658.1 | |
| GU727649.1 | 1 | 167 | ||
| U75586.1 | 2 | 269 | AAD09221.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3096n100 | Chr.17:8904185 - 9034134 on Build GRCh38 | Gain |
|
| esv25265 | Chr.17:9020861 - 9023559 on Build GRCh38 | Loss |
|
| nsv827882 | Chr.17:8979800 - 9039598 on Build GRCh38 | Gain |
|
| nsv833354 | Chr.17:8921846 - 9075675 on Build GRCh38 | Loss |
|
| nsv516433 | Chr.17:9017282 - 9023857 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map