Assay Details
Target Gene Details
Entrez Gene ID: | 10379 |
Gene Name: | interferon regulatory factor 9 |
Gene Aliases: |
IRF-9, ISGF3, ISGF3G, p48 |
Location: |
Chr.14:24161213-24166565 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRF9 | NM_006084.4 | 7 | 854 | NP_006075.3 |
| AK295467.1 | 6 | 1226 | ||
| BC035716.2 | 6 | 730 | AAH35716.1 | |
| M87503.1 | 7 | 761 | AAA58687.1 | |
| U92994.1 |
More Information
Set Membership: |
|

Genomic Map