Assay Details
Target Gene Details
Entrez Gene ID: | 2243 |
Gene Name: | fibrinogen alpha chain |
Gene Aliases: |
Fib2 |
Location: |
Chr.4:154583126-154590766 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGA | NM_000508.4 | 6 | 2148 | NP_000499.1 |
| BQ064506.1 | 2 | 585 | ||
| M58569.1 | 5 | 2802 | AAC97142.1 |
More Information
Set Membership: |
|

Genomic Map