Assay Details
Target Gene Details
Entrez Gene ID: | 10939 |
Gene Name: | AFG3 like matrix AAA peptidase subunit 2 |
Gene Aliases: |
SCA28, SPAX5 |
Location: |
Chr.18:12328944-12377309 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AFG3L2 | NM_006796.2 | 17 | 2673 | NP_006787.2 |
| XM_011525601.2 | 16 | 2505 | XP_011523903.1 | |
| AK129504.1 | 10 | 1499 | ||
| BC024282.1 | 18 | 2526 | AAH24282.1 | |
| BC065016.1 | 17 | 2595 | AAH65016.1 | |
| BU786821.1 | 1 | 546 | ||
| BX091241.1 | ||||
| Y18314.1 | 17 | 2592 | CAB48398.1 |
Target Gene Details
Entrez Gene ID: | 84617 |
Gene Name: | tubulin beta 6 class V |
Gene Aliases: |
HsT1601, TUBB-5 |
Location: |
Chr.18:12307669-12329826 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBB6 | NM_001303525.1 | 4 | 846 | NP_001290454.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060808 | Chr.18:12302793 - 12514418 on Build GRCh38 | Gain |
|
| nsv576496 | Chr.18:12247873 - 12490503 on Build GRCh38 | Gain |
|
| nsv1063125 | Chr.18:12046893 - 12918252 on Build GRCh38 | Loss |
|
| esv2758714 | Chr.18:12240682 - 12499526 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map