Assay Details
Target Gene Details
Entrez Gene ID: | 51475 |
Gene Name: | calcium binding protein 2 |
Gene Aliases: |
DFNB93 |
Location: |
Chr.11:67518947-67523428 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABP2 | NM_001318496.1 | NP_001305425.1 | ||
| NM_016366.2 | NP_057450.2 | |||
| AB593136.1 | ||||
| AF169154.1 | AAF25788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422195 | Chr.11:67466851 - 67859430 on Build GRCh38 | Duplication |
|
| dgv1977n54 | Chr.11:67397024 - 67521123 on Build GRCh38 | Loss |
|
| dgv52n111 | Chr.11:67362651 - 67521123 on Build GRCh38 | Deletion |
|
| nsv521723 | Chr.11:67408221 - 67543787 on Build GRCh38 | Loss |
|
| nsv527056 | Chr.11:67439955 - 67526837 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149124353] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map