Assay Details
Target Gene Details
Entrez Gene ID: | 27152 |
Gene Name: | inturned planar cell polarity protein |
Gene Aliases: |
INT, PDZD6, PDZK6 |
Location: |
Chr.4:127623271-127716779 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTU | NM_015693.3 | NP_056508.2 | ||
| XM_011531844.2 | XP_011530146.1 | |||
| XM_011531845.2 | XP_011530147.1 | |||
| XM_011531846.1 | XP_011530148.1 | |||
| XM_011531847.1 | XP_011530149.1 | |||
| XM_011531849.2 | XP_011530151.1 | |||
| XM_011531850.2 | XP_011530152.1 | |||
| XM_011531851.2 | XP_011530153.1 | |||
| XM_017008026.1 | XP_016863515.1 | |||
| AB033110.1 | ||||
| AK291481.1 | ||||
| BC051698.1 | AAH51698.1 | |||
| BC130611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830056 | Chr.4:127595984 - 127755402 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map