Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC4 | NM_001136022.2 | 10 | 4858 | NP_001129494.1 |
| NM_001198965.1 | 10 | 3763 | NP_001185894.1 | |
| NM_001198966.2 | 10 | 3741 | NP_001185895.1 | |
| NM_001198967.2 | 11 | 3775 | NP_001185896.1 | |
| NM_001288802.1 | 9 | 4649 | NP_001275731.1 | |
| NM_001320043.1 | 11 | 4099 | NP_001306972.1 | |
| NM_004554.4 | 10 | 4087 | NP_004545.2 | |
| BC053855.1 | 10 | 3903 | AAH53855.1 | |
| EU887634.1 | 10 | 4830 | ||
| EU887637.1 | 10 | 4646 | ||
| EU887640.1 | 10 | 4699 | ||
| EU887643.1 | 9 | 4525 | ||
| EU887646.1 | 9 | 4649 | ||
| EU887649.1 | 8 | 4771 | ||
| EU887652.1 | 6 | 3522 |
More Information
Set Membership: |
|

Genomic Map