Assay Details
Target Gene Details
Entrez Gene ID: | 55732 |
Gene Name: | chromosome 1 open reading frame 112 |
Gene Aliases: |
- |
Location: |
Chr.1:169794730-169854080 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf112 | NM_001320047.1 | NP_001306976.1 | ||
| NM_001320048.1 | NP_001306977.1 | |||
| NM_018186.3 | NP_060656.2 | |||
| XM_005245317.4 | XP_005245374.1 | |||
| XM_011509735.2 | XP_011508037.1 | |||
| XM_017001722.1 | XP_016857211.1 | |||
| XM_017001723.1 | XP_016857212.1 | |||
| XM_017001724.1 | XP_016857213.1 | |||
| XM_017001725.1 | XP_016857214.1 | |||
| AK001568.1 | BAA91761.1 | |||
| AK023532.1 | BAB14600.1 | |||
| AK096493.1 | ||||
| AK127098.1 | ||||
| AK294569.1 | ||||
| AK303296.1 | ||||
| AK310638.1 | ||||
| AL354614.1 | CAB89727.1 | |||
| BC091516.1 | AAH91516.1 | |||
| BC107168.1 | AAI07169.1 | |||
| BC107169.1 | AAI07170.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758976 | Chr.1:169812865 - 169916045 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142988055] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map