Assay Details
Target Gene Details
Entrez Gene ID: | 140738 |
Gene Name: | transmembrane protein 37 |
Gene Aliases: |
PR, PR1 |
Location: |
Chr.2:119429912-119438520 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM37 | NM_183240.2 | 2 | 493 | NP_899063.2 |
| XM_005263598.4 | 2 | 793 | XP_005263655.1 | |
| XM_006712300.3 | 2 | 617 | XP_006712363.1 | |
| XM_011510659.2 | 2 | 510 | XP_011508961.1 | |
| AF361356.1 | 2 | 459 | AAL50051.1 | |
| BC046362.1 | 1 | 618 | AAH46362.1 | |
| DR003856.1 | 2 | 500 | ||
| DR005637.1 | 2 | 482 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592167 | Chr.2:119401097 - 119522758 on Build GRCh38 | Gain |
|
| nsv459119 | Chr.2:118952824 - 119514207 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map