Assay Details
Target Gene Details
Entrez Gene ID: | 10652 |
Gene Name: | YKT6 v-SNARE homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.7:44200968-44214294 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YKT6 | NM_006555.3 | 7 | 1641 | NP_006546.1 |
| XM_005249582.4 | 8 | 1693 | XP_005249639.1 | |
| XM_017011682.1 | 6 | 1550 | XP_016867171.1 | |
| AK075018.1 | 2 | 990 | ||
| AK097209.1 | 1 | 1644 | ||
| BC007319.2 | 7 | 1571 | AAH07319.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map