Assay Details
Target Gene Details
Entrez Gene ID: | 1294 |
Gene Name: | collagen type VII alpha 1 |
Gene Aliases: |
EBD1, EBDCT, EBR1, NDNC8 |
Location: |
Chr.3:48564073-48595302 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 62 - Exon 63 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL7A1 | NM_000094.3 | NP_000085.1 | ||
| XM_011533337.1 | XP_011531639.1 | |||
| XM_017005688.1 | XP_016861177.1 | |||
| XM_017005689.1 | XP_016861178.1 | |||
| XM_017005690.1 | XP_016861179.1 | |||
| XM_017005691.1 | XP_016861180.1 | |||
| XM_017005692.1 | XP_016861181.1 | |||
| AB209645.1 | BAD92882.1 | |||
| L02870.1 | AAA75438.1 |
Target Gene Details
Entrez Gene ID: | 100313843 |
Gene Name: | microRNA 711 |
Gene Aliases: |
hsa-mir-711 |
Location: |
Chr.3:48578902-48578977 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR711 | NR_031756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954484 | Chr.3:48533268 - 48609567 on Build GRCh38 | Deletion |
|
| nsv1004383 | Chr.3:48506361 - 48612339 on Build GRCh38 | Loss |
|
| nsv3811 | Chr.3:48561747 - 48606376 on Build GRCh38 | Deletion |
|
| nsv834685 | Chr.3:48527576 - 48708341 on Build GRCh38 | Loss |
|
| nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss |
|
| nsv818138 | Chr.3:48573492 - 48694054 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114580573] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map