Assay Details
Target Gene Details
Entrez Gene ID: | 139728 |
Gene Name: | pregnancy up-regulated nonubiquitous CaM kinase |
Gene Aliases: |
BSTK3, CaMK1b |
Location: |
Chr.X:153669723-153687568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNCK | NM_001039582.3 | NP_001034671.3 | ||
| NM_001135740.1 | NP_001129212.1 | |||
| XM_005274651.2 | XP_005274708.1 | |||
| XM_006724809.3 | XP_006724872.1 | |||
| XM_011531107.2 | XP_011529409.2 | |||
| XM_011531108.2 | XP_011529410.1 | |||
| XM_011531110.2 | XP_011529412.1 | |||
| XM_011531111.2 | XP_011529413.1 | |||
| XM_011531112.2 | XP_011529414.1 | |||
| XM_017029277.1 | XP_016884766.1 | |||
| XM_017029278.1 | XP_016884767.1 | |||
| XM_017029279.1 | XP_016884768.1 | |||
| XM_017029280.1 | XP_016884769.1 | |||
| AK296067.1 | ||||
| AK296165.1 | ||||
| AK296203.1 | ||||
| AK303746.1 | ||||
| AK309148.1 | 4 | 782 | ||
| AK309159.1 | 2 | 266 | ||
| AK316402.1 | ||||
| BC064422.1 | 4 | 418 | AAH64422.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map