Assay Details
Target Gene Details
Entrez Gene ID: | 10913 |
Gene Name: | ectodysplasin A receptor |
Gene Aliases: |
DL, ECTD10A, ECTD10B, ED1R, ED3, ED5, EDA-A1R, EDA1R, EDA3, HRM1 |
Location: |
Chr.2:108894471-108989372 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDAR | NM_022336.3 | 12 | 3459 | NP_071731.1 |
| XM_006712204.1 | 11 | 3439 | XP_006712267.1 | |
| XM_011510502.2 | 11 | 3318 | XP_011508804.2 | |
| XM_011510503.2 | 12 | 3215 | XP_011508805.2 | |
| AF130988.1 | 12 | 3460 | AAD50076.1 | |
| CR749352.1 | 12 | 3568 | CAH18205.1 |
Target Gene Details
Entrez Gene ID: | 5903 |
Gene Name: | RAN binding protein 2 |
Gene Aliases: |
ADANE, ANE1, IIAE3, NUP358, TRP1, TRP2 |
Location: |
Chr.2:108719446-109842301 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RANBP2 | XM_017004623.1 | XP_016860112.1 |
More Information
Set Membership: |
|

Genomic Map