Assay Details
Target Gene Details
Entrez Gene ID: | 23385 |
Gene Name: | nicastrin |
Gene Aliases: |
ATAG1874 |
Location: |
Chr.1:160343056-160358952 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCSTN | NM_001290184.1 | NP_001277113.1 | ||
| NM_001290186.1 | NP_001277115.1 | |||
| NM_015331.2 | NP_056146.1 | |||
| XM_005245053.4 | XP_005245110.1 | |||
| XM_011509363.2 | XP_011507665.1 | |||
| AF240468.1 | AAG11412.1 | |||
| AK299142.1 | ||||
| AK310741.1 | ||||
| AK314764.1 | ||||
| AY359120.1 | AAQ89478.1 | |||
| BC047621.1 | AAH47621.1 | |||
| D87442.1 | BAA13383.1 |
More Information
Set Membership: |
|

Genomic Map