Assay Details
Target Gene Details
Entrez Gene ID: | 5806 |
Gene Name: | pentraxin 3 |
Gene Aliases: |
TNFAIP5, TSG-14 |
Location: |
Chr.3:157436791-157443628 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTX3 | NM_002852.3 | 3 | 1312 | NP_002843.2 |
| AK312705.1 | ||||
| BC039733.1 | 3 | 1285 | AAH39733.1 | |
| BQ010272.1 | ||||
| M31166.1 | 3 | 1242 | AAA61234.1 | |
| X63053.1 | 3 | 1174 | CAA44778.1 | |
| X63613.1 | 3 | 1236 | CAA45158.1 |
Target Gene Details
Entrez Gene ID: | 79674 |
Gene Name: | ventricular zone expressed PH domain containing 1 |
Gene Aliases: |
MELT, VEPH |
Location: |
Chr.3:157259742-157503626 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VEPH1 | NM_001167911.1 | NP_001161383.1 | ||
| NM_001167912.1 | NP_001161384.1 | |||
| NM_024621.2 | NP_078897.2 | |||
| XM_011513134.2 | XP_011511436.1 | |||
| XM_011513135.1 | XP_011511437.1 | |||
| AB051479.1 | ||||
| AK027625.1 | BAB55243.1 | |||
| AL713656.1 | CAD28465.2 | |||
| BC101660.1 | AAI01661.1 | |||
| BC111017.1 | AAI11018.1 | |||
| BC113555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759193 | Chr.3:157372377 - 157536895 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map