Assay Details
Target Gene Details
Entrez Gene ID: | 50855 |
Gene Name: | par-6 family cell polarity regulator alpha |
Gene Aliases: |
PAR-6A, PAR6, PAR6C, PAR6alpha, TAX40, TIP-40 |
Location: |
Chr.16:67660948-67662778 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARD6A | NM_001037281.1 | 3 | 535 | NP_001032358.1 |
| NM_016948.2 | 3 | 538 | NP_058644.1 | |
| XM_005255977.3 | 3 | 563 | XP_005256034.1 | |
| XM_011523095.2 | 3 | 560 | XP_011521397.1 | |
| XM_011523096.2 | 3 | 542 | XP_011521398.1 | |
| XM_017023261.1 | 3 | 506 | XP_016878750.1 | |
| AB041642.1 | 3 | 500 | BAB16105.1 | |
| AB043634.1 | 3 | 527 | BAA96235.1 | |
| AF028827.1 | 1 | 104 | AAB84252.1 | |
| AF252292.1 | 3 | 576 | AAF71529.1 | |
| AF265565.1 | 3 | 509 | AAF75548.1 | |
| AJ277095.1 | 3 | 498 | CAB85490.1 | |
| BC015626.2 | 3 | 515 | AAH15626.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv952046 | Chr.16:67628798 - 67678697 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map