Assay Details
Target Gene Details
Entrez Gene ID: | 6563 |
Gene Name: | solute carrier family 14 member 1 (Kidd blood group) |
Gene Aliases: |
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE |
Location: |
Chr.18:45724123-45752520 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC14A1 | NM_001128588.3 | 1 | 109 | NP_001122060.3 |
| NM_001146036.2 | 1 | 109 | NP_001139508.2 | |
| NM_001308278.1 | 1 | 113 | NP_001295207.1 | |
| NM_001308279.1 | 1 | 113 | NP_001295208.1 | |
| NM_015865.6 | 1 | 109 | NP_056949.4 | |
| XM_005258329.1 | 1 | 85 | XP_005258386.1 | |
| XM_005258333.1 | 1 | 83 | XP_005258390.1 | |
| XM_006722526.3 | 1 | 109 | XP_006722589.1 | |
| XM_011526144.1 | 1 | 85 | XP_011524446.1 | |
| AF328892.1 | ||||
| AK127452.1 | ||||
| AK289608.1 | ||||
| AK294129.1 | ||||
| AK295268.1 | ||||
| BC050539.1 | AAH50539.1 | |||
| DA134800.1 | 1 | 109 | ||
| DA156178.1 | 1 | 113 | ||
| DA186606.1 | 1 | 85 | ||
| DA473671.1 | ||||
| U35735.1 | AAB00181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063292 | Chr.18:45715129 - 45737040 on Build GRCh38 | Gain |
|
| nsv525268 | Chr.18:45168400 - 45894830 on Build GRCh38 | Gain |
|
| esv3893100 | Chr.18:45542509 - 45858191 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map