Assay Details
Target Gene Details
Entrez Gene ID: | 55225 |
Gene Name: | ribonucleoprotein, PTB binding 2 |
Gene Aliases: |
- |
Location: |
Chr.1:64744979-64833232 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAVER2 | NM_018211.3 | 3 | 519 | NP_060681.2 |
| XM_006710738.3 | 3 | 626 | XP_006710801.2 | |
| XM_011541706.2 | 3 | 635 | XP_011540008.1 | |
| AB046799.1 | 3 | 519 |
More Information
Set Membership: |
|

Genomic Map