Assay Details
Target Gene Details
Entrez Gene ID: | 10370 |
Gene Name: | Cbp/p300 interacting transactivator with Glu/Asp rich carboxy-terminal domain 2 |
Gene Aliases: |
ASD8, MRG-1, MRG1, P35SRJ, VSD2 |
Location: |
Chr.6:139372255-139374650 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CITED2 | NM_001168388.2 | 2 | 1578 | NP_001161860.1 |
| NM_001168389.2 | 2 | 1561 | NP_001161861.2 | |
| NM_006079.4 | 2 | 1717 | NP_006070.2 | |
| AA400897.1 | 1 | 221 | ||
| AA767624.1 | ||||
| AF109161.1 | 2 | 1687 | AAD10055.1 | |
| AI087283.1 | 1 | 225 | ||
| BC004377.1 | 2 | 1670 | AAH04377.1 | |
| CF126189.1 | 1 | 509 |
More Information
Set Membership: |
|

Genomic Map