Assay Details
Target Gene Details
Entrez Gene ID: | 6948 |
Gene Name: | transcobalamin 2 |
Gene Aliases: |
D22S676, D22S750, II, TC, TC II, TC-2, TC2, TCII |
Location: |
Chr.22:30607083-30627060 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCN2 | NM_000355.3 | NP_000346.2 | ||
| NM_001184726.1 | NP_001171655.1 | |||
| AK225445.1 | ||||
| AK313711.1 | ||||
| BC001176.1 | AAH01176.1 | |||
| BC011239.1 | AAH11239.1 | |||
| CR456591.1 | CAG30477.1 | |||
| L02647.1 | AAA61056.1 | |||
| L02648.1 | AAA61057.1 | |||
| M60396.1 | AAA61054.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834175 | Chr.22:30580637 - 30693558 on Build GRCh38 | Gain |
|
| nsv829171 | Chr.22:30202041 - 30647591 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map