Assay Details
Target Gene Details
Entrez Gene ID: | 128821 |
Gene Name: | cystatin 9-like |
Gene Aliases: |
CTES7B, bA218C14.1 |
Location: |
Chr.20:23564732-23568749 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CST9L | NM_080610.2 | 1 | 471 | NP_542177.1 |
| AK312112.1 | 1 | 469 | ||
| AY358615.1 | 1 | 414 | AAQ88978.1 | |
| BC029656.1 | 1 | 471 | AAH29656.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585723 | Chr.20:23446011 - 23588763 on Build GRCh38 | Gain |
|
| esv3645525 | Chr.20:23425607 - 23597079 on Build GRCh38 | Gain |
|
| nsv1065390 | Chr.20:23432253 - 23588601 on Build GRCh38 | Loss |
|
| nsv470544 | Chr.20:23444001 - 23588763 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113667569] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map