Assay Details
Target Gene Details
Entrez Gene ID: | 128239 |
Gene Name: | IQ motif containing GTPase activating protein 3 |
Gene Aliases: |
- |
Location: |
Chr.1:156525405-156572604 on Build GRCh38 |
Assay Gene Location: | Within Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQGAP3 | NM_178229.4 | 38 | 5262 | NP_839943.2 |
| XM_011509198.2 | 38 | 5251 | XP_011507500.1 | |
| XM_011509200.2 | 27 | 4431 | XP_011507502.1 | |
| XM_011509201.2 | 26 | 3820 | XP_011507503.1 | |
| XM_017000317.1 | 38 | 5248 | XP_016855806.1 | |
| XM_017000318.1 | 36 | 5005 | XP_016855807.1 | |
| AB105103.1 | 38 | 5198 | BAC78211.1 | |
| AK122653.1 | 24 | 3470 | BAC85501.1 | |
| BC131536.1 | ||||
| BM910515.1 | 4 | 739 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952215 | Chr.1:156524609 - 156527308 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map