Assay Details
Target Gene Details
Entrez Gene ID: | 26507 |
Gene Name: | cyclin and CBS domain divalent metal cation transport mediator 1 |
Gene Aliases: |
ACDP1, CLP-1 |
Location: |
Chr.10:99329099-99394330 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNNM1 | NM_020348.2 | 1 | 1368 | NP_065081.2 |
| XM_005269692.3 | 1 | 1368 | XP_005269749.2 | |
| XM_011539629.1 | 1 | 1368 | XP_011537931.1 | |
| XM_011539630.1 | 1 | 1368 | XP_011537932.1 | |
| XM_011539631.2 | 1 | 1368 | XP_011537933.1 | |
| AF169226.1 | 1 | 191 | AAF86357.1 | |
| AK296545.1 | 1 | 536 | ||
| AK299346.1 | 1 | 525 | ||
| AK308618.1 | 1 | 1109 | ||
| BC098103.1 | 1 | 142 | ||
| BC098134.1 | 1 | 142 | AAH98134.1 | |
| BC098279.1 | 1 | 142 | ||
| BC098307.1 | 1 | 142 | ||
| DA758649.1 | 1 | 525 |
More Information
Set Membership: |
|

Genomic Map