Assay Details
Target Gene Details
Entrez Gene ID: | 2274 |
Gene Name: | four and a half LIM domains 2 |
Gene Aliases: |
AAG11, DRAL, FHL-2, SLIM-3, SLIM3 |
Location: |
Chr.2:105360826-105438773 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FHL2 | NM_001039492.2 | 6 | 854 | NP_001034581.1 |
| NM_001318894.1 | 5 | 1100 | NP_001305823.1 | |
| NM_001318895.1 | 6 | 1088 | NP_001305824.1 | |
| NM_001318896.1 | 5 | 803 | NP_001305825.1 | |
| NM_001318897.1 | 4 | 854 | NP_001305826.1 | |
| NM_001318898.1 | 5 | 913 | NP_001305827.1 | |
| NM_001318899.1 | 7 | 976 | NP_001305828.1 | |
| NM_001450.3 | 5 | 1037 | NP_001441.4 | |
| NM_201555.1 | 7 | 1209 | NP_963849.1 | |
| NM_201557.3 | 5 | 1029 | NP_963851.2 | |
| XM_011510798.2 | 6 | 836 | XP_011509100.1 | |
| AB158503.1 | 4 | 576 | BAD69710.1 | |
| AK313841.1 | 6 | 886 | ||
| AL521653.3 | 5 | 730 | ||
| AY634685.1 | 4 | 630 | AAV35468.1 | |
| BC012742.2 | 7 | 1209 | AAH12742.1 | |
| BC014397.1 | 7 | 1209 | AAH14397.1 | |
| BC093049.1 | 5 | 795 | ||
| BI092736.1 | 2 | 147 | ||
| BI562043.1 | 5 | 690 | ||
| BM559077.1 | 7 | 858 | ||
| BT006960.1 | 4 | 630 | AAP35606.1 | |
| BX340370.2 | ||||
| CF552311.1 | 5 | 760 | ||
| CR457437.1 | 4 | 630 | CAG33718.1 | |
| CR936682.1 | 5 | 1100 | ||
| CR936799.1 | 5 | 6081 | ||
| DQ266432.1 | 6 | 1088 | ABB73038.1 | |
| DQ307067.1 | 5 | 967 | ABC25549.1 | |
| JQ313902.1 | 4 | 600 | ||
| L42176.1 | 6 | 768 | AAA85333.1 | |
| U29332.1 | 5 | 751 | AAC52073.1 | |
| U60117.1 | 2 | 253 | AAC50794.1 |
More Information
Set Membership: |
|

Genomic Map