Assay Details
Target Gene Details
Entrez Gene ID: | 102 |
Gene Name: | ADAM metallopeptidase domain 10 |
Gene Aliases: |
AD10, AD18, CD156c, CDw156, HsT18717, MADM, RAK, kuz |
Location: |
Chr.15:58595204-58749978 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAM10 | NM_001110.3 | 16 | 3098 | NP_001101.1 |
| NM_001320570.1 | 15 | 3005 | NP_001307499.1 | |
| XM_017021861.1 | 17 | 3153 | XP_016877350.1 | |
| AF009615.1 | 16 | 3123 | AAC51766.1 | |
| AK023460.1 | ||||
| BX372217.2 | 1 | 862 |
More Information
Additional Information:
For this assay, SNP(s) [rs77874096] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map