Assay Details
Target Gene Details
Entrez Gene ID: | 619208 |
Gene Name: | family with sequence similarity 229 member B |
Gene Aliases: |
C6orf225 |
Location: |
Chr.6:112087471-112102790 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM229B | NM_001033564.2 | NP_001028736.1 | ||
| XM_017011174.1 | XP_016866663.1 | |||
| XM_017011175.1 | XP_016866664.1 | |||
| AL049461.1 | ||||
| BC044912.2 | AAH44912.1 | |||
| BF110308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3571360 | Chr.6:112095330 - 112101319 on Build GRCh38 | Loss |
|

Genomic Map