Assay Details
Target Gene Details
Entrez Gene ID: | 54954 |
Gene Name: | family with sequence similarity 120C |
Gene Aliases: |
CXorf17, ORF34 |
Location: |
Chr.X:54068324-54183281 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM120C | NM_001300788.1 | NP_001287717.1 | ||
| NM_017848.5 | 13 | 2777 | NP_060318.4 | |
| XM_006724589.3 | 13 | 2773 | XP_006724652.1 | |
| AY150025.1 | 13 | 2777 | AAO24121.1 | |
| BC136413.1 | 13 | 2774 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map