Assay Details
Target Gene Details
Entrez Gene ID: | 6548 |
Gene Name: | solute carrier family 9 member A1 |
Gene Aliases: |
APNH, LIKNS, NHE-1, NHE1, PPP1R143 |
Location: |
Chr.1:27098809-27155130 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A1 | NM_003047.4 | 12 | 4160 | NP_003038.2 |
| NR_046474.1 | 11 | 3699 | ||
| XM_011542021.2 | 13 | 3238 | XP_011540323.1 | |
| AK124218.1 | 7 | 2589 | ||
| BQ775239.1 | 1 | 598 | ||
| BQ775318.1 | 1 | 598 | ||
| M81768.1 | 12 | 3417 | AAB59460.1 | |
| M96066.1 | ||||
| M96067.1 | 8 | 1897 | ||
| S68616.1 | 12 | 3941 | AAC60606.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834535 | Chr.1:27006585 - 27179926 on Build GRCh38 | Gain+Loss |
|
| nsv1000012 | Chr.1:26714969 - 27141528 on Build GRCh38 | Gain |
|
| nsv509058 | Chr.1:27068848 - 27109365 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map