Assay Details
Target Gene Details
Entrez Gene ID: | 59 |
Gene Name: | actin, alpha 2, smooth muscle, aorta |
Gene Aliases: |
AAT6, ACTSA, MYMY5 |
Location: |
Chr.10:88935074-88991397 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTA2 | NM_001141945.2 | NP_001135417.1 | ||
| NM_001320855.1 | NP_001307784.1 | |||
| NM_001613.2 | NP_001604.1 | |||
| AK300353.1 | ||||
| AK300664.1 | ||||
| AK313294.1 | ||||
| AY692464.1 | ||||
| BC017554.2 | AAH17554.1 | |||
| BC093052.1 | AAH93052.1 | |||
| BP248338.1 | ||||
| BP379410.1 | ||||
| DC304647.1 | ||||
| DC354672.1 | ||||
| DC359878.1 | ||||
| DC385559.1 | ||||
| J05192.1 | AAA51577.1 | |||
| X13839.1 | CAA32064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831943 | Chr.10:88905079 - 89106077 on Build GRCh38 | Gain |
|
| nsv831942 | Chr.10:88804940 - 88970595 on Build GRCh38 | Gain |
|
| esv3624144 | Chr.10:88876937 - 89011636 on Build GRCh38 | Gain |
|
| esv3624139 | Chr.10:88724980 - 89003307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map