Assay Details
Target Gene Details
Entrez Gene ID: | 10629 |
Gene Name: | TATA-box binding protein associated factor 6 like |
Gene Aliases: |
PAF65A |
Location: |
Chr.11:62771303-62787342 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF6L | NM_006473.3 | NP_006464.1 | ||
| XM_005273714.1 | XP_005273771.1 | |||
| AF069735.1 | AAC39905.1 | |||
| AK289499.1 | ||||
| AK310054.1 | ||||
| AK314330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555175 | Chr.11:62771357 - 62787614 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map