Assay Details
Target Gene Details
Entrez Gene ID: | 23096 |
Gene Name: | IQ motif and Sec7 domain 2 |
Gene Aliases: |
BRAG1, MRX1, MRX78 |
Location: |
Chr.X:53225784-53321350 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQSEC2 | NM_001111125.2 | 15 | 5223 | NP_001104595.1 |
| NM_015075.1 | 14 | 4450 | NP_055890.1 | |
| XM_006724579.2 | 16 | 5345 | XP_006724642.1 | |
| XM_006724580.3 | 15 | 4513 | XP_006724643.1 | |
| XM_006724581.3 | XP_006724644.1 | |||
| XM_006724582.3 | XP_006724645.1 | |||
| XM_006724583.3 | XP_006724646.1 | |||
| XM_011530773.2 | 14 | 4417 | XP_011529075.1 | |
| XM_017029359.1 | 14 | 5219 | XP_016884848.1 | |
| XM_017029360.1 | 15 | 4631 | XP_016884849.1 | |
| AB011094.1 | 15 | 5223 | BAA25448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2740175 | Chr.X:52862926 - 55652533 on Build GRCh38 | Deletion |
|
| nsv435891 | Chr.X:52861979 - 55653313 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map