Assay Details
Target Gene Details
Entrez Gene ID: | 815 |
Gene Name: | calcium/calmodulin dependent protein kinase II alpha |
Gene Aliases: |
CAMKA |
Location: |
Chr.5:150219491-150289840 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK2A | NM_015981.3 | 19 | 4596 | NP_057065.2 |
| NM_171825.2 | 18 | 4563 | NP_741960.1 | |
| AB023185.1 | 18 | 4496 | ||
| BC040457.1 | 18 | 4478 | AAH40457.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763509 | Chr.5:150215547 - 150245974 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map