Assay Details
Target Gene Details
Entrez Gene ID: | 3592 |
Gene Name: | interleukin 12A |
Gene Aliases: |
CLMF, IL-12A, NFSK, NKSF1, P35 |
Location: |
Chr.3:159988836-159996019 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12A | NM_000882.3 | NP_000873.2 | ||
| AF101062.1 | AAD16432.1 | |||
| AF180562.1 | AAD56385.1 | |||
| BC104982.1 | AAI04983.1 | |||
| BC104984.1 | AAI04985.1 | |||
| M65271.1 | AAA35694.1 | |||
| M65291.1 | AAA59937.1 |
Target Gene Details
Entrez Gene ID: | 101928376 |
Gene Name: | IL12A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:159913400-160207092 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12A-AS1 | NR_108088.1 | |||
| AK097161.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829769 | Chr.3:159889124 - 160087495 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map