Assay Details
Target Gene Details
Entrez Gene ID: | 26056 |
Gene Name: | RAB11 family interacting protein 5 |
Gene Aliases: |
GAF1, RIP11, pp75 |
Location: |
Chr.2:73073382-73113018 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAB11FIP5 | NM_015470.2 | NP_056285.1 | ||
| XM_005264251.3 | XP_005264308.1 | |||
| XM_005264252.3 | XP_005264309.1 | |||
| XM_006711985.3 | XP_006712048.1 | |||
| XM_011532753.2 | XP_011531055.1 | |||
| XM_017003788.1 | XP_016859277.1 | |||
| AB020664.1 | ||||
| AF153085.1 | AAF34356.1 | |||
| AF334812.1 | AAK20892.1 | |||
| AK023823.1 | ||||
| AL117579.1 | CAB56002.1 | |||
| BC035013.1 | AAH35013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002295 | Chr.2:72989026 - 73123263 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map