Assay Details
Target Gene Details
Entrez Gene ID: | 90806 |
Gene Name: | angel homolog 2 |
Gene Aliases: |
Ccr4d, KIAA0759L |
Location: |
Chr.1:212992182-213015875 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGEL2 | NM_001300753.1 | 8 | 1552 | NP_001287682.1 |
| NM_001300755.1 | 8 | 1650 | NP_001287684.1 | |
| NM_001300757.1 | 7 | 1305 | NP_001287686.1 | |
| NM_001300758.1 | 7 | 1301 | NP_001287687.1 | |
| NM_144567.4 | 8 | 1627 | NP_653168.2 | |
| NR_125333.1 | 6 | 1174 | ||
| XM_005273344.1 | 8 | 1648 | XP_005273401.1 | |
| XM_005273345.1 | 8 | 1694 | XP_005273402.1 | |
| XM_005273346.2 | 8 | 1447 | XP_005273403.1 | |
| XM_005273347.3 | 7 | 1319 | XP_005273404.1 | |
| XM_017002774.1 | 8 | 1576 | XP_016858263.1 | |
| XM_017002775.1 | 9 | 1875 | XP_016858264.1 | |
| XM_017002776.1 | 9 | 1899 | XP_016858265.1 | |
| XM_017002777.1 | 9 | 1945 | XP_016858266.1 | |
| XM_017002778.1 | 9 | 1688 | XP_016858267.1 | |
| AF510741.1 | 7 | 1256 | AAM44053.1 | |
| AK093947.1 | 3 | 2051 | ||
| AK294479.1 | 8 | 1426 | ||
| AK295116.1 | 8 | 1556 | ||
| AK299208.1 | 8 | 1620 | ||
| AK299365.1 | 6 | 1084 | ||
| AK310946.1 | 6 | 793 | ||
| AL079275.1 | 8 | 1585 | ||
| BC047469.1 | 7 | 1217 | AAH47469.1 |
More Information
Set Membership: |
|

Genomic Map