Assay Details
Target Gene Details
Entrez Gene ID: | 8531 |
Gene Name: | Y-box binding protein 3 |
Gene Aliases: |
CSDA, CSDA1, DBPA, ZONAB |
Location: |
Chr.12:10699077-10723448 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YBX3 | NM_001145426.1 | NP_001138898.1 | ||
| NM_003651.4 | NP_003642.3 | |||
| XM_011520871.2 | XP_011519173.1 | |||
| XM_017020122.1 | XP_016875611.1 | |||
| XM_017020123.1 | XP_016875612.1 | |||
| AB209896.1 | BAD93133.1 | |||
| AK314846.1 | ||||
| AU117252.1 | ||||
| BC008801.2 | AAH08801.1 | |||
| BC009744.2 | AAH09744.1 | |||
| BC015564.1 | AAH15564.1 | |||
| BC015913.1 | AAH15913.1 | |||
| BC021926.1 | AAH21926.1 | |||
| M24069.1 | AAA35749.1 | |||
| X72712.1 | CAA51261.1 | |||
| X95325.1 | CAA64631.1 |
More Information
Set Membership: |
|

Genomic Map