Assay Details
Target Gene Details
Entrez Gene ID: | 5019 |
Gene Name: | 3-oxoacid CoA-transferase 1 |
Gene Aliases: |
OXCT, SCOT |
Location: |
Chr.5:41730065-41870689 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXCT1 | NM_000436.3 | 1 | 358 | NP_000427.1 |
| XM_017009494.1 | 1 | 239 | XP_016864983.1 | |
| AK312327.1 | 1 | 54 | ||
| BC009001.1 | 1 | 82 | AAH09001.1 | |
| BE244429.1 | 1 | 173 | ||
| CR457049.1 | CAG33330.1 | |||
| U62961.1 | 1 | 125 | AAB07366.1 |
Target Gene Details
Entrez Gene ID: | 100874002 |
Gene Name: | OXCT1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:41870030-41872241 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OXCT1-AS1 | NR_046635.1 | |||
| DB470733.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4811 | Chr.5:41844420 - 41887251 on Build GRCh38 | Deletion |
|
| nsv830273 | Chr.5:41867194 - 42065968 on Build GRCh38 | Gain |
|
| esv3604851 | Chr.5:41591779 - 41992753 on Build GRCh38 | Gain |
|
| nsv830271 | Chr.5:41716425 - 41895608 on Build GRCh38 | Loss |
|
| nsv830272 | Chr.5:41833263 - 41986065 on Build GRCh38 | Gain |
|
| dgv1096e214 | Chr.5:41866397 - 41966396 on Build GRCh38 | Gain |
|
| esv2762518 | Chr.5:41780108 - 41935011 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map