Assay Details
Target Gene Details
Entrez Gene ID: | 57650 |
Gene Name: | KIAA1524 |
Gene Aliases: |
CIP2A, p90 |
Location: |
Chr.3:108545752-108589644 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KIAA1524 | NM_020890.2 | 9 | 1331 | NP_065941.2 |
| XM_006713716.3 | 9 | 1115 | XP_006713779.1 | |
| XM_006713717.2 | 10 | 1227 | XP_006713780.1 | |
| XM_011513056.1 | 9 | 1075 | XP_011511358.1 | |
| XM_011513057.2 | 10 | 1196 | XP_011511359.1 | |
| AB040957.1 | 9 | 1058 | BAA96048.1 | |
| AF334474.1 | 9 | 1130 | AAL11632.1 | |
| AK056933.1 | 9 | 1140 | ||
| AK308315.1 | 9 | 1077 | ||
| AK310446.1 | 9 | 1335 | ||
| AL713689.1 | 9 | 1126 | CAD28489.1 | |
| BC063433.1 | 9 | 1122 | AAH63433.1 | |
| BC130564.1 | 9 | 1098 | ||
| BC136371.1 | 9 | 1098 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524016 | Chr.3:108566875 - 108589473 on Build GRCh38 | Loss |
|
| nsv829658 | Chr.3:108484408 - 108666131 on Build GRCh38 | Gain |
|
| nsv1011158 | Chr.3:107746970 - 108629120 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map