Assay Details
Target Gene Details
Entrez Gene ID: | 2712 |
Gene Name: | glycerol kinase 2 |
Gene Aliases: |
GKP2, GKTA |
Location: |
Chr.4:79406353-79408218 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GK2 | NM_033214.2 | NP_149991.2 | ||
| AK292241.1 | ||||
| BC029820.1 | AAH29820.1 | |||
| BC048274.1 | ||||
| BC058888.1 | AAH58888.1 | |||
| X78712.1 | CAA55365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999174 | Chr.4:79291781 - 79591202 on Build GRCh38 | Loss |
|
| esv2727859 | Chr.4:78716995 - 79557290 on Build GRCh38 | Deletion |
|
| nsv1006026 | Chr.4:78111837 - 79966019 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map