Assay Details
Target Gene Details
Entrez Gene ID: | 142689 |
Gene Name: | ankyrin repeat and SOCS box containing 12 |
Gene Aliases: |
- |
Location: |
Chr.X:64224192-64230631 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB12 | NM_130388.3 | 2 | 241 | NP_569059.3 |
| AF403030.1 | AAL57349.1 | |||
| AK096896.1 | 2 | 221 | BAC04888.1 | |
| AK300684.1 | 2 | 217 | ||
| BC069436.1 | AAH69436.1 | |||
| BC069555.1 | AAH69555.1 | |||
| BC093883.1 | 1 | 82 | AAH93883.1 | |
| BC104946.1 | 1 | 82 | AAI04947.1 | |
| DA560184.1 | 2 | 243 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521560 | Chr.X:64063724 - 64245972 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map