Assay Details
Target Gene Details
Entrez Gene ID: | 25790 |
Gene Name: | cilia and flagella associated protein 45 |
Gene Aliases: |
CCDC19, NESG1 |
Location: |
Chr.1:159872364-159900116 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP45 | NM_012337.2 | NP_036469.2 | ||
| AF094758.1 | AAD55817.1 | |||
| AK225566.1 | ||||
| AK225582.1 | ||||
| AK292249.1 | ||||
| AK308262.1 | ||||
| BC089391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|

Genomic Map