Assay Details
Target Gene Details
Entrez Gene ID: | 5706 |
Gene Name: | proteasome 26S subunit, ATPase 6 |
Gene Aliases: |
CADP44, HEL-S-73, P44, SUG2, p42 |
Location: |
Chr.14:52707163-52727998 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMC6 | NM_002806.3 | NP_002797.3 | ||
| XM_017021472.1 | XP_016876961.1 | |||
| AF006305.1 | AAB61616.1 | |||
| AK299155.1 | ||||
| AK313670.1 | ||||
| BC005390.1 | AAH05390.1 | |||
| BM788029.1 | ||||
| BT006843.1 | AAP35489.1 | |||
| CR456709.1 | CAG32990.1 | |||
| D78275.1 | BAA11338.1 | |||
| FJ224299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832796 | Chr.14:52680158 - 52860182 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map