Assay Details
Target Gene Details
Entrez Gene ID: | 51028 |
Gene Name: | vacuolar protein sorting 36 homolog |
Gene Aliases: |
C13orf9, CGI-145, EAP45 |
Location: |
Chr.13:52412602-52450678 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS36 | NM_001282168.1 | 14 | 3903 | NP_001269097.1 |
| NM_001282169.1 | 14 | 4064 | NP_001269098.1 | |
| NM_016075.3 | 14 | 3930 | NP_057159.2 | |
| AK022911.1 | 1 | 2017 | ||
| AK023182.1 | 12 | 3647 | BAB14451.1 | |
| BC037279.1 | 14 | 3879 | AAH37279.1 | |
| BX647847.1 | 14 | 3871 | ||
| CR933653.1 | CAI45953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428288 | Chr.13:52154235 - 52493997 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map