Assay Details
Target Gene Details
Entrez Gene ID: | 80777 |
Gene Name: | cytochrome b5 type B |
Gene Aliases: |
CYB5-M, CYPB5M, OMB5 |
Location: |
Chr.16:69424595-69466264 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYB5B | NM_030579.2 | 5 | 1769 | NP_085056.2 |
| BC004373.1 | 5 | 1704 | AAH04373.1 | |
| BC014431.1 | 5 | 1727 | AAH14431.2 |
More Information
Set Membership: |
|

Genomic Map